Pre-printed ID tapes
ID tape, printed with 'Quarantine', White/red, -, 50 mm
Catalog # 300-0561
Supplier: Sampling Systems
Jump to:
Product Details & Documents
ID tape helps to prevent handling errors by allowing containers, pallets, bags and so on to be easily labelled with highly visible messaging. Operators can quickly identify the status of a container to prevent it from being used in error.
- Use with standard tape dispensers
- Medium strength adhesion
ID tape is an ideal way of labelling large or small items. These tape rolls come pre-printed with 'Quarantine', 'Sampled', 'Released' or 'Rejected'.
Packaging: 10 rolls per box.
Specifications
- Sterile:-
- Width:50 mm
- Colour:White/red
- Description:ID tape, printed with 'Quarantine'
- Seq:01