BAECHLER DSC HIGH PRESSURE GOLD-PLATED CRUCIBLE WITHOUT LOCATING PIN,WITH LID AND SEAL HEIGHT 5.9 MM, VOLUME 50 UL,PRESSURE TIGHTNES S WITH WATER 200 BAR (MAX 217 BAR) MAXIMUM TEMPERATURE 400 DEGREE CELSIU
Catalog # MSPP116031
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together