0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Volumetric pipette with ring mark, conformity certified, 20 ml 1 * 1 items
Catalog # HECH42556064
Supplier:
GLASWARENFABRIK KARL HECHT
Expand image
Volumetric pipette with ring mark, conformity certified, 20 ml 1 * 1 items
Catalog # HECH42556064
Supplier:
GLASWARENFABRIK KARL HECHT
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection