ContactUsLinkComponent
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures. Ideal Use: 0.2ml PCR plates or tubes Holes: 96 (8 x 12) Hole Depth: 16mm Hole Diam.: 6.4mm Dimensions: 127mm x 85mm x 25mm 1 * 1 items
Catalog # DIVECHAM-A100
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures. Ideal Use: 0.2ml PCR plates or tubes Holes: 96 (8 x 12) Hole Depth: 16mm Hole Diam.: 6.4mm Dimensions: 127mm x 85mm x 25mm 1 * 1 items
Catalog # DIVECHAM-A100
Frequently Bought Together