ContactUsLinkComponent
Recombinat Protein,Human TFPI Protein, His Tag,Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Asp 29 - Lys 282 (Accession NP006278.1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 50 µG
Catalog # ACRBTFI-H5226-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TFPI Protein, His Tag,Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Asp 29 - Lys 282 (Accession NP006278.1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 50 µG
Catalog # ACRBTFI-H5226-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together