ContactUsLinkComponent
ActiveMax® Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P 13232). 1 * 20 µG
Catalog # ACRBIL7-H4219-20UG
Supplier:  ACRO Biosystems
ActiveMax® Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P 13232). 1 * 20 µG
Catalog # ACRBIL7-H4219-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together