ContactUsLinkComponent
Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C- 1 * 1 mg
Catalog # ACRBNE4-C52H4-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Cynomolgus Nectin-4 Protein, His Tag,Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells ( HEK293). It contains AA Gly 32 - Ser 349 (Accession XP005541277.1).This protein carries a polyhistidine tag at the C- 1 * 1 mg
Catalog # ACRBNE4-C52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together