ContactUsLinkComponent
Recombinat Protein,Mouse TSLP Protein, His Tag,Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contai ns AA Tyr 20 - Glu 140 (Accession Q9JIE6-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBTSP-M52H8-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse TSLP Protein, His Tag,Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contai ns AA Tyr 20 - Glu 140 (Accession Q9JIE6-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBTSP-M52H8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together