ContactUsLinkComponent
Human Nectin-1, Fc Tag (PV1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession Q15223-1). 1 * 1 mg
Catalog # ACRBPV1-H5253-1MG
Supplier:  ACRO Biosystems
Human Nectin-1, Fc Tag (PV1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession Q15223-1). 1 * 1 mg
Catalog # ACRBPV1-H5253-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together