ContactUsLinkComponent
Human HGF R, His Tag (MET-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession AAI30421.1). 1 * 1 mg
Catalog # ACRBMET-H5227-1MG
Supplier:  ACRO Biosystems
Human HGF R, His Tag (MET-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession AAI30421.1). 1 * 1 mg
Catalog # ACRBMET-H5227-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together