ContactUsLinkComponent
Cynomolgus Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acce ssion G7N0K6-1). 1 * 1 mg
Catalog # ACRBFG1-C5269-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acce ssion G7N0K6-1). 1 * 1 mg
Catalog # ACRBFG1-C5269-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together