ContactUsLinkComponent
Recombinat Protein,Human Cystatin D CST5 Protein, His Tag,Human Cystatin D, His Tag (CS5-H5220) is expressed from human 293 cells (H EK293). It contains AA Gly 21 - Val 142 (Accession AAH69514).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Catalog # ACRBCS5-H5220-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Cystatin D CST5 Protein, His Tag,Human Cystatin D, His Tag (CS5-H5220) is expressed from human 293 cells (H EK293). It contains AA Gly 21 - Val 142 (Accession AAH69514).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Catalog # ACRBCS5-H5220-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together