ContactUsLinkComponent
ActiveMax® Human Flt-3 Ligand, Tag Free (FLL-H5218) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Acc ession AAI26294). 1 * 1 mg
Catalog # ACRBFLL-H5218-1MG
Supplier:  ACRO Biosystems
ActiveMax® Human Flt-3 Ligand, Tag Free (FLL-H5218) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Acc ession AAI26294). 1 * 1 mg
Catalog # ACRBFLL-H5218-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together