ContactUsLinkComponent
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 1 mg
Catalog # ACRBGMF-H4214-1MG
Supplier:  ACRO Biosystems
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 1 mg
Catalog # ACRBGMF-H4214-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together