ContactUsLinkComponent
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1). 1 * 1 mg
Catalog # ACRBER4-H5221-1MG
Supplier:  ACRO Biosystems
Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession NP005226.1). 1 * 1 mg
Catalog # ACRBER4-H5221-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together