ContactUsLinkComponent
Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession Q15303-1). 1 * 1 mg
Catalog # ACRBER4-H5251-1MG
Supplier:  ACRO Biosystems
Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession Q15303-1). 1 * 1 mg
Catalog # ACRBER4-H5251-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together