ContactUsLinkComponent
MABSol® Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Access ion NP942088). 1 * 25 µG
  ACRBCD7-H82F6-25UG
MABSol® Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Access ion NP942088). 1 * 25 µG
  ACRBCD7-H82F6-25UG
Frequently Bought Together