ContactUsLinkComponent
Cynomolgus Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acc ession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus 1 * 1 mg
Catalog # ACRBCD7-C52H1-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acc ession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus 1 * 1 mg
Catalog # ACRBCD7-C52H1-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together