ContactUsLinkComponent
MABSol® Biotinylated Mouse PCSK9, Avitag,His Tag (PC9-M82E1) is expressed from human HEK293 cells. It contains AA Gln 35 - Gln 694 ( Accession AAH38085.1). 1 * 25 µG
Catalog # ACRBPC9-M82E1-25UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Mouse PCSK9, Avitag,His Tag (PC9-M82E1) is expressed from human HEK293 cells. It contains AA Gln 35 - Gln 694 ( Accession AAH38085.1). 1 * 25 µG
Catalog # ACRBPC9-M82E1-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together