ContactUsLinkComponent
Biotinylated Human CD23, His,Avitag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accessi on P06734-1). 1 * 25 µG
Catalog # ACRBCD3-H82Q5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD23, His,Avitag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accessi on P06734-1). 1 * 25 µG
Catalog # ACRBCD3-H82Q5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together