ContactUsLinkComponent
Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 100 µG
  ACRB2B4-H5255100UG
Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 100 µG
  ACRB2B4-H5255100UG
Frequently Bought Together