Recombinat Protein,Human ALCAM Protein, Fc Tag,Human ALCAM, Fc Tag (CD6-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Trp 28 - Ala 526 (Accession NP001618).This protein carries a human IgG1 Fc tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBCD6-H5259-1MG
Supplier: ACRO Biosystems
Frequently Bought Together