ContactUsLinkComponent
Recombinat Protein,Biotinylated Human EMMPRIN CD147 Protein, Avitag™,His Tag,Biotinylated Human EMMPRIN, Avitag,His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession P35613-2).This protein carries 1 * 200 µG
Catalog # ACRBCD7-H82E0200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human EMMPRIN CD147 Protein, Avitag™,His Tag,Biotinylated Human EMMPRIN, Avitag,His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession P35613-2).This protein carries 1 * 200 µG
Catalog # ACRBCD7-H82E0200UG
Supplier:  ACRO Biosystems
Frequently Bought Together