ContactUsLinkComponent
Recombinat Protein,Human Angiopoietin-2 ANGPT2 (275-496) Protein, His Tag,Human Angiopoietin-2 (275-496), His Tag (AN2-H5242) is exp ressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This protein carries a 1 * 100 µG
Catalog # ACRBAN2-H5242100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-2 ANGPT2 (275-496) Protein, His Tag,Human Angiopoietin-2 (275-496), His Tag (AN2-H5242) is exp ressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This protein carries a 1 * 100 µG
Catalog # ACRBAN2-H5242100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together