ContactUsLinkComponent
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are 1 * 200 µG
  ACRBBCA-C82F4200UG
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are 1 * 200 µG
  ACRBBCA-C82F4200UG
Frequently Bought Together