ContactUsLinkComponent
Recombinat Protein,Human BACE-1 Protein, His Tag,Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It co ntains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBBA1-H5220-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human BACE-1 Protein, His Tag,Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It co ntains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBBA1-H5220-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together