Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H522 6) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein carr 1 * 1 mg
Catalog # ACRBCA9-H5226-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together