ContactUsLinkComponent
Biotinylated Mouse BAFFR, Fc,Avitag (BAR-M82F0) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accessio n NP082351). 1 * 25 µG
  ACRBBAR-M82F0-25UG
Biotinylated Mouse BAFFR, Fc,Avitag (BAR-M82F0) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accessio n NP082351). 1 * 25 µG
  ACRBBAR-M82F0-25UG
Frequently Bought Together