ContactUsLinkComponent
Biotinylated Human CD64, His,Avitag (SPR  BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 200 µG
  ACRBFCA-H82E8200UG
Biotinylated Human CD64, His,Avitag (SPR  BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 200 µG
  ACRBFCA-H82E8200UG
Frequently Bought Together