ContactUsLinkComponent
Biotinylated Human IL-4 R alpha, Avitag,His Tag (ILR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP000409.1). 1 * 200 µG
Catalog # ACRBILR-H82E9200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-4 R alpha, Avitag,His Tag (ILR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP000409.1). 1 * 200 µG
Catalog # ACRBILR-H82E9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together