ContactUsLinkComponent
Biotinylated Human Cynomolgus Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhesus 1 * 200 µG
Catalog # ACRBCD8-H82F2200UG
Supplier:  ACRO Biosystems
Biotinylated Human Cynomolgus Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhesus 1 * 200 µG
Catalog # ACRBCD8-H82F2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together