ContactUsLinkComponent
Cynomolgus Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contain s AA Ala 20 - Arg 191 (Accession XP005569963.1). In the region Ala 20 - Arg 191, the AA sequence of Cynomolgus and Rh 1 * 1 mg
Catalog # ACRBCD7-C5259-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque CD27, Mouse IgG2a Fc Tag, low endotoxin (CD7-C5259) is expressed from human 293 cells (HEK293). It contain s AA Ala 20 - Arg 191 (Accession XP005569963.1). In the region Ala 20 - Arg 191, the AA sequence of Cynomolgus and Rh 1 * 1 mg
Catalog # ACRBCD7-C5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together