ContactUsLinkComponent
Biotinylated Human BTLA (31-134), Fc,Avitag (BTA-H82F8) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession Q7Z6A9). 1 * 25 µG
Catalog # ACRBBTA-H82F8-25UG
Supplier:  ACRO Biosystems
Biotinylated Human BTLA (31-134), Fc,Avitag (BTA-H82F8) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession Q7Z6A9). 1 * 25 µG
Catalog # ACRBBTA-H82F8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together