ContactUsLinkComponent
Recombinat Protein,Human CD94 Protein, His Tag,Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contai ns AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD4-H52H8100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD94 Protein, His Tag,Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contai ns AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD4-H52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together