ContactUsLinkComponent
Mouse PD-L1, Fc Tag (PD1-M5251) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession NP068693). 1 * 200 µG
Catalog # ACRBPD1-M5251200UG
Supplier:  ACRO Biosystems
Mouse PD-L1, Fc Tag (PD1-M5251) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession NP068693). 1 * 200 µG
Catalog # ACRBPD1-M5251200UG
Supplier:  ACRO Biosystems
Frequently Bought Together