ContactUsLinkComponent
Rat PD-1, Fc Tag (PD1-R5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP001100397.1). 1 * 100 µG
  ACRBPD1-R5253100UG
Marketsource
Rat PD-1, Fc Tag (PD1-R5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP001100397.1). 1 * 100 µG
  ACRBPD1-R5253100UG
Marketsource
Frequently Bought Together