ContactUsLinkComponent
Recombinat Protein,Rat LAG3 CD223 Protein, His Tag,Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It cont ains AA Gly 24 - Leu 442 (Accession NP997678).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBLA3-R52H5-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Rat LAG3 CD223 Protein, His Tag,Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It cont ains AA Gly 24 - Leu 442 (Accession NP997678).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBLA3-R52H5-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together