ContactUsLinkComponent
Mouse B7-1, Fc Tag (CD0-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession Q00609-1). 1 * 1 mg
Catalog # ACRBCD0-M5259-1MG
Supplier:  ACRO Biosystems
Mouse B7-1, Fc Tag (CD0-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession Q00609-1). 1 * 1 mg
Catalog # ACRBCD0-M5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together