ContactUsLinkComponent
Recombinat Protein,Mouse ADAM17 TACE CD156b Protein, His Tag,Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (H EK293). It contains AA Pro 18 - Asp 563 (Accession Q9Z0F8).This protein carries a polyhistidine tag at the C-terminus 1 * 500 µG
Catalog # ACRBAD7-M52H1500UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse ADAM17 TACE CD156b Protein, His Tag,Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (H EK293). It contains AA Pro 18 - Asp 563 (Accession Q9Z0F8).This protein carries a polyhistidine tag at the C-terminus 1 * 500 µG
Catalog # ACRBAD7-M52H1500UG
Supplier:  ACRO Biosystems
Frequently Bought Together