ContactUsLinkComponent
Recombinat Protein,Rhesus macaque EGF R Protein, His Tag,Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P55245).This protein carries a polyhistidine tag at the C-termi 1 * 50 µG
Catalog # ACRBEGR-C52H1-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque EGF R Protein, His Tag,Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P55245).This protein carries a polyhistidine tag at the C-termi 1 * 50 µG
Catalog # ACRBEGR-C52H1-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together