ContactUsLinkComponent
Biotinylated Human B7-H3 (4Ig), His,Avitag (B7B-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 ( Accession Q5ZPR3-1). 1 * 25 µG
Catalog # ACRBB7B-H82E8-25UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H3 (4Ig), His,Avitag (B7B-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 ( Accession Q5ZPR3-1). 1 * 25 µG
Catalog # ACRBB7B-H82E8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together