Mouse FCGRTB2M Heterodimer Protein, His TagTwin-Strep Tag (BLI verified) (FCM-M52W2) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q61559-1 (FCGRT) P01887-1 (B2M)). 1 * 1 mg
Catalog # ACRBFCM-M52W2-1MG
Supplier: ACRO Biosystems
Frequently Bought Together