ContactUsLinkComponent
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
Catalog # ACRBGRA-H52H7-1MG
Supplier:  ACRO Biosystems
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
Catalog # ACRBGRA-H52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together