ContactUsLinkComponent
Rhesus macaque OX40, Fc Tag (OX0-C5251) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Ala 214 (Accession XP001 090870.1). 1 * 100 µG
  ACRBOX0-C5251100UG
Rhesus macaque OX40, Fc Tag (OX0-C5251) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Ala 214 (Accession XP001 090870.1). 1 * 100 µG
  ACRBOX0-C5251100UG
Frequently Bought Together