ContactUsLinkComponent
Recombinat Protein,Human TIMP-2 Protein, His Tag,Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It co ntains AA Cys 27 - Pro 220 (Accession NP003246.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 50 µG
Catalog # ACRBTI2-H5223-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TIMP-2 Protein, His Tag,Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It co ntains AA Cys 27 - Pro 220 (Accession NP003246.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 50 µG
Catalog # ACRBTI2-H5223-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together