Mouse recombinant PLGF (from HEK293)
50 µG
ACRBPGF-M52H0-50UG
Marketsource
- Pk:50 µG
- Protease-free:Y
- Protein/peptide name:PLGF
- Protein synonyms:PGF|PLGF|PlGF2|PGFL|PlGF|SHGC-10760
- Purity:>95%
- Endotoxin level:Low
- Source:HEK293 cells
- Biological activity:Lyophilised from 0,22 μm filtered solution in 100 mM HAC, pH 2,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. Immobilised Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) at 2 μg/ml (100 μl/well) can bind Mouse PLGF, His Tag (Cat. No. ACRBPGF-M52H0) with a linear range of 0,2 - 4 ng/ml (QC tested).
- Molecular weight:17,8 kDa
- Shipping temperature:Powder, RT
- Nuclease-free:Y
- Cat. No.:ACRBPGF-M52H0-50UG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Storage conditions:–20 °C
- Gene ID:P49764-1
- Carrier-free:Y
- Formulation:Lyophilised from 0,22 μm filtered solution in 100 mM HAC, pH 2,5. Normally trehalose is added as protectant before lyophilisation
- Conjugation:Unconjugated
- Species:Mouse
- Animal-free:Y



