Biotinylated Human ANGPTL7, Mouse IgG2a Fc,Avitag (AN7-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pr o 346 (Accession O43827-1). 1 * 25 µG
Catalog # ACRBAN7-H82F3-25UG
Supplier: ACRO Biosystems
Frequently Bought Together