Human recombinant complement factor D (from HEK293 cells)
50 µG
Catalog # ACRBCFD-H52H8-50UG
Marketsource
Supplier: ACRO Biosystems
Human recombinant complement factor D (from HEK293 cells)
Catalog # ACRBCFD-H52H8-50UG
Supplier: ACRO Biosystems
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Human Complement Factor D / CFD Protein, His Tag (active enzyme), ACROBiosystems.
Specifications
Specifications
- Pk:50 µG
- Protein/peptide name:Complement Factor D
- Protein synonyms:PFD|Complement factor D|CFD|DF|Adipsin
- Purity:>90% as determined by SDS-PAGE.
- Endotoxin level:Low
- Source:HEK293 cells
- Molecular weight:26,3 kDa
- Shipping temperature:RT
- Cat. No.:ACRBCFD-H52H8-50UG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Storage conditions:–20 °C
- Gene ID:P00746-1
- Formulation:Lyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.
- Conjugation:Unconjugated
- Tag sequence:His Tag (active enzyme)
- Species:Human



