Auftragserfassung
Switzerland
ContactUsLinkComponent
Human CD47, His Tag, low endotoxin (CD7-H522a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessio n NP942088). 1 * 50 µG
Catalog # ACRBCD7-H522A-50UG
Supplier:  ACRO Biosystems
Human CD47, His Tag, low endotoxin (CD7-H522a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessio n NP942088). 1 * 50 µG
Catalog # ACRBCD7-H522A-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together