ContactUsLinkComponent
Recombinat Protein,Human NKp30 NCR3 CD337 Protein, His Tag,Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK29 3). It contains AA Leu 19 - Gly 135 (Accession AAH52582).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Catalog # ACRBNC3-H5228100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human NKp30 NCR3 CD337 Protein, His Tag,Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK29 3). It contains AA Leu 19 - Gly 135 (Accession AAH52582).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Catalog # ACRBNC3-H5228100UG
Supplier:  ACRO Biosystems
Frequently Bought Together